Assay Details
Gene Symbol: KNL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004941298.2 | XP_004941355.1 | 8 - 9 | 517 | 74 | |
XM_015286271.1 | XP_015141757.1 | 8 - 9 | 520 | 74 | ||
XM_420938.5 | XP_420938.4 | 8 - 9 | 520 | 74 | ||
GenBank mRNA | BX934082.2 | - | 8 - 9 | 427 | 74 | |
BX934169.1 | - | 6 - 7 | 269 | 74 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
