Assay Details
Gene Symbol: RIDA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001234688.5 | XP_001234689.4 | 2 - 3 | 201 | 81 | |
GenBank mRNA | BX932754.1 | - | 2 - 3 | 193 | 81 | |
BX935048.1 | - | 2 - 3 | 175 | 81 |

More Information
Set Membership: |
![]() ![]() ![]() |
