Assay Details
Gene Symbol: COX16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001197057.1 | NP_001183986.1 | 2 - 3 | 190 | 134 | |
GenBank mRNA | BX935937.1 | - | 2 - 3 | 144 | 134 |

More Information
Set Membership: |
![]() ![]() ![]() |
