Assay Details
Gene Symbol: ORMDL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001231892.5 | XP_001231893.1 | 2 - 3 | 384 | 66 | |
XM_015289355.2 | XP_015144841.1 | 2 - 3 | 286 | 66 | ||
XM_025152270.1 | XP_025008038.1 | 2 - 3 | 313 | 66 | ||
GenBank mRNA | CR352957.1 | - | 1 - 2 | 74 | 66 | |
CR523921.1 | - | 2 - 3 | 386 | 66 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
