Assay Details
Gene Symbol: COX16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001197057.1 | NP_001183986.1 | 1 - 2 | 108 | 94 | |
GenBank mRNA | BX932336.1 | - | 1 - 2 | 63 | 94 | |
CR353996.1 | - | 1 - 2 | 65 | 94 | ||
CR405829.1 | - | 1 - 2 | 44 | 94 |

More Information
Set Membership: |
![]() ![]() ![]() |
