Assay Details
Gene Symbol: POLR2D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001277409.1 | NP_001264338.1 | 1 - 2 | 98 | 291 | |
GenBank mRNA | BX930879.2 | - | 1 - 2 | 79 | 126 | |
BX933730.2 | - | 1 - 2 | 69 | 291 | ||
CR390815.1 | - | 1 - 2 | 79 | 291 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
