Assay Details
Gene Symbol: C1QTNF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001277702.1 | NP_001264631.1 | 1 - 2 | 128 | 62 | |
GenBank mRNA | BX930999.2 | - | 1 - 2 | 128 | 62 |

More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001277702.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
