Assay Details
Gene Symbol: C1QTNF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001277702.1 | NP_001264631.1 | 2 - 3 | 386 | 76 | |
GenBank mRNA | BX930999.2 | - | 2 - 3 | 386 | 76 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
