Assay Details
Gene Symbol: PEBP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001198642.1 | NP_001185571.1 | 2 - 3 | 303 | 61 | |
GenBank mRNA | BX933434.2 | - | 2 - 3 | 264 | 61 | |
BX934603.1 | - | 2 - 3 | 287 | 61 | ||
CN222635.1 | - | 2 - 3 | 303 | 61 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
