Assay Details
Gene Symbol: TFPI2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015281937.2 | XP_015137423.1 | 2 - 3 | 324 | 63 | |
XM_418662.6 | XP_418662.2 | 2 - 3 | 324 | 63 | ||
GenBank mRNA | BX934121.2 | - | 2 - 3 | 314 | 63 | |
BX950344.1 | - | 2 - 3 | 309 | 63 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
