Assay Details
Gene Symbol: RAMP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_025152394.1 | XP_025008162.1 | 1 - 2 | 199 | 75 | |
GenBank mRNA | BX929334.1 | - | 1 - 2 | 106 | 75 | |
BX936098.2 | - | 1 - 2 | 117 | 75 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
