Assay Details
Gene Symbol: PRRG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001277736.1 | NP_001264665.1 | 1 - 2 | 103 | 76 | |
XM_015301833.2 | XP_015157319.1 | 1 - 2 | 137 | 76 | ||
GenBank mRNA | BX935305.1 | - | 1 - 2 | 103 | 76 | |
BX936218.2 | - | 1 - 2 | 103 | 76 |

More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001277736.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
