Assay Details
Gene Symbol: MRPL46 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001277112.1 | NP_001264041.1 | 1 - 2 | 254 | 89 | |
GenBank mRNA | BX930828.2 | - | 1 - 2 | 205 | 89 | |
BX930890.1 | - | 1 - 2 | 186 | 89 | ||
BX935417.1 | - | 1 - 2 | 227 | 89 | ||
BX950417.2 | - | 1 - 2 | 254 | 89 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
