Assay Details
Gene Symbol: MYOZ1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004942083.3 | XP_004942140.1 | 2 - 3 | 348 | 69 | |
XM_421619.5 | XP_421619.3 | 3 - 4 | 470 | 69 | ||
GenBank mRNA | BX935806.1 | - | 2 - 3 | 348 | 69 | |
BX935870.1 | - | 1 - 2 | 88 | 69 | ||
BX950467.2 | - | 1 - 2 | 88 | 69 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
