Assay Details
Gene Symbol: ETNK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015291728.2 | XP_015147214.1 | 2 - 3 | 423 | 81 | |
XM_416426.6 | XP_416426.3 | 1 - 2 | 229 | 81 | ||
GenBank mRNA | CR385222.1 | - | 1 - 2 | 218 | 81 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
