Assay Details
Gene Symbol: GAS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001199470.1 | NP_001186399.1 | 3 - 4 | 457 | 69 | |
XM_015286176.2 | XP_015141662.1 | 4 - 5 | 812 | 69 | ||
XM_015286177.2 | XP_015141663.1 | 4 - 5 | 888 | 69 | ||
XM_015286178.2 | XP_015141664.1 | 4 - 5 | 578 | 69 | ||
XM_015286179.2 | XP_015141665.1 | 4 - 5 | 805 | 69 | ||
GenBank mRNA | CR385836.1 | - | 4 - 5 | 576 | 69 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
