Assay Details
Gene Symbol: MRPL23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001199385.1 | NP_001186314.1 | 2 - 3 | 152 | 140 | |
GenBank mRNA | AB101640.1 | - | 2 - 3 | 159 | 143 | |
BX933781.1 | - | 1 - 2 | 213 | 140 | ||
CO503845.1 | - | 2 - 3 | 190 | 140 |

More Information
Set Membership: |
![]() ![]() ![]() |
