Assay Details
Gene Symbol: BAG3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015288906.2 | XP_015144392.1 | 1 - 2 | 201 | 95 | |
XM_015288907.2 | XP_015144393.1 | 1 - 2 | 201 | 95 | ||
XM_015288908.2 | XP_015144394.1 | 1 - 2 | 201 | 95 | ||
GenBank mRNA | AJ720653.1 | - | 1 - 2 | 200 | 95 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
