Assay Details
Gene Symbol: AADAC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004943567.3 | XP_004943624.1 | 4 - 5 | 2273 | 118 | |
XM_015291844.1 | XP_015147330.1 | 5 - 6 | 2428 | 118 | ||
XM_025153460.1 | XP_025009228.1 | 4 - 5 | 2273 | 118 | ||
XM_025153461.1 | XP_025009229.1 | 4 - 5 | 2269 | 118 | ||
XM_422836.6 | XP_422836.2 | 4 - 5 | 664 | 118 | ||
GenBank mRNA | BX929777.1 | - | 4 - 5 | 620 | 118 | |
BX931204.1 | - | 4 - 5 | 654 | 118 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
