Assay Details
Gene Symbol: CENPV | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_415846.6 | XP_415846.2 | 3 - 4 | 562 | 81 | |
GenBank mRNA | CR352613.1 | - | 3 - 4 | 400 | 81 | |
CR389739.1 | - | 3 - 4 | 410 | 81 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
