Assay Details
Gene Symbol: ACOT13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015282414.2 | XP_015137900.1 | 2 - 3 | 329 | 115 | |
GenBank mRNA | BX929282.2 | - | 2 - 3 | 284 | 115 | |
BX932290.1 | - | 2 - 3 | 317 | 115 | ||
BX935853.1 | - | 2 - 3 | 293 | 115 | ||
CR406050.1 | - | 2 - 3 | 295 | 115 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
