Assay Details
Gene Symbol: ZNF469 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004944285.3 | XP_004944342.1 | 4 | 8236 | 107 | |
XM_004944286.3 | XP_004944343.1 | 6 | 7783 | 107 | ||
XM_025154329.1 | XP_025010097.1 | 3 | 8057 | 107 | ||
GenBank mRNA | CR406896.1 | - | 1 | 652 | 107 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
