Assay Details
Gene Symbol: PRPS1L1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_420181.6 | XP_420181.1 | 3 - 4 | 579 | 97 | |
GenBank mRNA | CR407240.1 | - | 3 - 4 | 573 | 97 |

More Information
Set Membership: |
![]() ![]() ![]() |
