Assay Details
Gene Symbol: BBX | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004938282.3 | XP_004938339.3 | 12 - 13 | 2707 | 66 | |
XM_015297349.2 | XP_015152835.2 | 13 - 14 | 3572 | 66 | ||
XM_015297371.2 | XP_015152857.2 | 12 - 13 | 3492 | 66 | ||
XM_025144174.1 | XP_024999942.1 | 13 - 14 | 3573 | 66 | ||
XM_025144175.1 | XP_024999943.1 | 13 - 14 | 3499 | 66 | ||
XM_025144177.1 | XP_024999945.1 | 12 - 13 | 3493 | 66 | ||
XM_025144178.1 | XP_024999946.1 | 11 - 12 | 3412 | 66 | ||
XM_025144181.1 | XP_024999949.1 | 12 - 13 | 3402 | 66 | ||
GenBank mRNA | CR524052.1 | - | 2 - 3 | 209 | 66 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
