Assay Details
Gene Symbol: REL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001167726.1 | NP_001161198.1 | 4 - 5 | 503 | 136 | |
XM_015283288.2 | XP_015138774.2 | 4 - 5 | 421 | 136 | ||
GenBank mRNA | AJ445799.1 | - | 4 - 5 | 504 | 136 | |
M26381.1 | - | 3 - 4 | 382 | 136 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
