Assay Details
Gene Symbol: REL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001167726.1 | NP_001161198.1 | 6 - 7 | 747 | 95 | |
XM_015283288.2 | XP_015138774.2 | 6 - 7 | 665 | 95 | ||
GenBank mRNA | M26381.1 | - | 5 - 6 | 626 | 95 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
