Assay Details
Gene Symbol: REL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001167726.1 | NP_001161198.1 | 7 - 8 | 953 | 91 | |
XM_015283288.2 | XP_015138774.2 | 7 - 8 | 871 | 91 | ||
GenBank mRNA | M26381.1 | - | 6 - 7 | 832 | 91 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
