Assay Details
Gene Symbol: BET1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001199678.1 | NP_001186607.1 | 1 - 2 | 145 | 149 | |
GenBank mRNA | BX930441.1 | - | 1 - 2 | 128 | 149 | |
BX932361.2 | - | 1 - 2 | 145 | 149 | ||
BX932727.1 | - | 1 - 2 | 136 | 149 |

More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001199678.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
