Assay Details
Gene Symbol: BET1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001199678.1 | NP_001186607.1 | 3 - 4 | 315 | 131 | |
GenBank mRNA | BX930441.1 | - | 3 - 4 | 298 | 131 | |
BX932361.2 | - | 3 - 4 | 315 | 131 | ||
BX932727.1 | - | 3 - 4 | 306 | 131 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
