Assay Details
Gene Symbol: PINLYP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_424499.6 | XP_424499.4 | 2 - 3 | 274 | 67 | |
GenBank mRNA | BX930636.1 | - | 2 - 3 | 225 | 67 | |
BX932148.1 | - | 2 - 3 | 261 | 67 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
