Assay Details
Gene Symbol: MALL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001199428.1 | NP_001186357.1 | 3 - 4 | 741 | 75 | |
GenBank mRNA | BU298453.1 | - | 3 - 4 | 426 | 75 | |
BX929438.2 | - | 3 - 4 | 742 | 75 | ||
BX930937.1 | - | 3 - 4 | 671 | 75 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
