Assay Details
Gene Symbol: CYTL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_420795.4 | XP_420795.1 | 1 - 2 | 327 | 137 | |
GenBank mRNA | BX931297.1 | - | 1 - 2 | 317 | 137 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
