Assay Details
Gene Symbol: CYTL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_420795.4 | XP_420795.1 | 3 - 4 | 497 | 107 | |
GenBank mRNA | BX931297.1 | - | 3 - 4 | 487 | 107 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
