Assay Details
Gene Symbol: IKBIP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001232182.5 | XP_001232183.1 | 1 - 2 | 233 | 132 | |
XM_416166.6 | XP_416166.1 | 1 - 2 | 233 | 132 | ||
GenBank mRNA | BX930962.1 | - | 1 - 2 | 212 | 133 | |
BX931370.2 | - | 1 - 2 | 267 | 133 | ||
BX931418.1 | - | 1 - 2 | 170 | 133 | ||
BX932424.1 | - | 1 - 2 | 118 | 133 | ||
BX932445.2 | - | 1 - 2 | 118 | 133 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_001232182.5, XM_416166.6 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
