Assay Details
Gene Symbol: IKBIP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_416166.6 | XP_416166.1 | 3 - 4 | 570 | 89 | |
GenBank mRNA | BX931370.2 | - | 3 - 4 | 605 | 89 | |
BX931418.1 | - | 3 - 4 | 508 | 89 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
