Assay Details
Gene Symbol: AAED1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003643010.4 | XP_003643058.1 | 5 - 6 | 3074 | 111 | |
GenBank mRNA | BX932659.1 | - | 4 - 5 | 517 | 111 |
Gene Symbol: CDC14B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004949233.3 | XP_004949290.1 | 1 - 2 | 1526 | 111 | |
XM_004949234.3 | XP_004949291.1 | 3 - 4 | 1122 | 111 | ||
XM_004949235.3 | XP_004949292.1 | 3 - 4 | 1122 | 111 | ||
XM_004949236.3 | XP_004949293.1 | 3 - 4 | 1122 | 111 | ||
XM_004949237.3 | XP_004949294.1 | 3 - 4 | 1122 | 111 | ||
XM_015280388.2 | XP_015135874.1 | 3 - 4 | 1122 | 111 | ||
XM_025144747.1 | XP_025000515.1 | 3 - 4 | 1122 | 111 | ||
XM_025144748.1 | XP_025000516.1 | 2 - 3 | 1315 | 111 | ||
XM_025144749.1 | XP_025000517.1 | 2 - 3 | 346 | 111 | ||
XM_025144750.1 | XP_025000518.1 | 3 - 4 | 1122 | 111 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_004949233.3, XM_004949234.3, XM_004949235.3, XM_004949236.3, XM_004949237.3, XM_015280388.2, XM_025144747.1, XM_025144748.1, XM_025144749.1, XM_025144750.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
