Assay Details
Gene Symbol: GNA13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_415686.6 | XP_415686.1 | 2 - 3 | 569 | 112 | |
GenBank mRNA | BX933165.1 | - | 2 - 3 | 515 | 112 |

More Information
Set Membership: |
![]() ![]() ![]() |
