Assay Details
Gene Symbol: PDZK1IP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001277593.1 | NP_001264522.1 | 1 - 2 | 144 | 78 | |
GenBank mRNA | BX933371.1 | - | 1 - 2 | 144 | 78 | |
CD727791.1 | - | 1 - 2 | 126 | 78 |

More Information
Set Membership: |
![]() ![]() ![]() |
