Assay Details
Gene Symbol: PDZK1IP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001277593.1 | NP_001264522.1 | 3 - 4 | 349 | 111 | |
GenBank mRNA | BX277854.2 | - | 2 - 3 | 122 | 111 | |
BX933371.1 | - | 3 - 4 | 349 | 111 | ||
CD727791.1 | - | 3 - 4 | 331 | 111 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
