Assay Details
Gene Symbol: PODN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004936802.3 | XP_004936859.1 | 2 - 3 | 1159 | 74 | |
XM_015291207.2 | XP_015146693.1 | 2 - 3 | 1257 | 74 | ||
XM_015291208.2 | XP_015146694.1 | 2 - 3 | 670 | 74 | ||
XM_025152988.1 | XP_025008756.1 | 4 - 5 | 5665 | 74 | ||
XM_025152989.1 | XP_025008757.1 | 4 - 5 | 5593 | 74 | ||
XM_025152990.1 | XP_025008758.1 | 4 - 5 | 6060 | 74 | ||
XM_025152991.1 | XP_025008759.1 | 2 - 3 | 5651 | 74 | ||
GenBank mRNA | BX933561.1 | - | 2 - 3 | 297 | 74 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
