Assay Details
Gene Symbol: ATP23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001199554.1 | NP_001186483.1 | 1 - 2 | 204 | 61 | |
XM_025147807.1 | XP_025003575.1 | 1 - 2 | 249 | 61 | ||
XM_025147823.1 | XP_025003591.1 | 1 - 2 | 168 | 61 | ||
GenBank mRNA | BU356502.1 | - | 1 - 2 | 177 | 61 | |
BU409185.1 | - | 1 - 2 | 232 | 61 | ||
CR352492.1 | - | 1 - 2 | 207 | 61 | ||
CR353901.1 | - | 1 - 2 | 166 | 61 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_025147823.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
