Assay Details
Gene Symbol: ATP23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001199554.1 | NP_001186483.1 | 2 - 3 | 254 | 82 | |
XM_025147807.1 | XP_025003575.1 | 2 - 3 | 299 | 82 | ||
XM_025147823.1 | XP_025003591.1 | 2 - 3 | 218 | 82 | ||
GenBank mRNA | BU356502.1 | - | 2 - 3 | 227 | 82 | |
BU409185.1 | - | 2 - 3 | 282 | 82 | ||
CR352492.1 | - | 2 - 3 | 257 | 82 | ||
CR353901.1 | - | 2 - 3 | 216 | 82 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_025147823.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
