Assay Details
Gene Symbol: TSPO2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_418037.5 | XP_418037.2 | 129 | 66 | ||
GenBank mRNA | BX931454.2 | - | 70 | 66 | ||
BX936041.2 | - | 61 | 66 | |||
CR353324.1 | - | 70 | 66 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_418037.5 transcripts.
Set Membership: |
![]() ![]() |
