Assay Details
Gene Symbol: TSPO2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004948410.3 | XP_004948467.1 | 64 | |||
XM_418037.5 | XP_418037.2 | 64 | ||||
GenBank mRNA | BX931454.2 | - | 289 | 64 | ||
BX934347.2 | - | 64 | ||||
BX936041.2 | - | 64 | ||||
CR353324.1 | - | 64 |
