Assay Details
Gene Symbol: PRTFDC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001199585.1 | NP_001186514.1 | 5 - 6 | 423 | 124 | |
XM_015281378.2 | XP_015136864.1 | 5 - 6 | 599 | 124 | ||
GenBank mRNA | BX932120.2 | - | 5 - 6 | 423 | 124 |

More Information
Set Membership: |
![]() ![]() ![]() |
