Assay Details
Gene Symbol: PLBD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015289354.2 | XP_015144840.2 | 6 - 7 | 911 | 77 | |
XM_416206.6 | XP_416206.5 | 6 - 7 | 911 | 77 | ||
GenBank mRNA | BX932425.2 | - | 1 - 2 | 43 | 77 | |
BX933306.2 | - | 6 - 7 | 849 | 77 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
