Assay Details
Gene Symbol: MRPS25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001277840.1 | NP_001264769.1 | 3 - 4 | 416 | 143 | |
GenBank mRNA | BX933213.2 | - | 3 - 4 | 377 | 143 | |
CN231166.1 | - | 3 - 4 | 417 | 144 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
