Assay Details
Gene Symbol: DTX3L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015289955.2 | XP_015145441.1 | 3 - 4 | 1958 | 118 | |
GenBank mRNA | BX950444.1 | - | 1 - 2 | 569 | 118 |

More Information
Set Membership: |
![]() ![]() ![]() |
