Assay Details
Gene Symbol: RAC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001201452.1 | NP_001188381.1 | 3 - 4 | 401 | 58 | |
GenBank mRNA | AJ398007.1 | - | 3 - 4 | 212 | 58 | |
AJ851813.1 | - | 3 - 4 | 365 | 58 | ||
BU358555.1 | - | 3 - 4 | 401 | 58 | ||
CF074925.1 | - | 3 - 4 | 347 | 58 | ||
CR353911.1 | - | 3 - 4 | 401 | 58 |

More Information
Set Membership: |
![]() ![]() ![]() |
