Assay Details
Gene Symbol: C1H3ORF38 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004938306.3 | XP_004938363.1 | 2 - 3 | 486 | 76 | |
XM_416640.6 | XP_416640.1 | 2 - 3 | 441 | 76 | ||
GenBank mRNA | BX929498.2 | - | 1 - 2 | 137 | 76 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
